Assay Details
Gene Symbol: Fgf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231437.2 | XP_006231499.1 | 2 - 3 | 69 | 189 | |
XM_008760398.1 | XP_008758620.1 | 2 - 3 | 69 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
