Assay Details
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012571.2 | NP_036703.2 | 1 - 2 | 205 | 64 | |
GenBank mRNA | BC061877.1 | - | 1 - 2 | 178 | 64 | |
CK600227.1 | - | 1 - 2 | 224 | 64 | ||
D00252.1 | - | 1 - 2 | 162 | 64 | ||
FM055803.1 | - | 1 - 2 | 127 | 64 | ||
J04171.1 | - | 1 - 2 | 146 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
