Assay Details
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012571.2 | NP_036703.2 | 2 - 3 | 386 | 93 | |
GenBank mRNA | BC061877.1 | - | 2 - 3 | 359 | 93 | |
CK600227.1 | - | 2 - 3 | 405 | 93 | ||
D00252.1 | - | 2 - 3 | 343 | 93 | ||
FM055803.1 | - | 2 - 3 | 308 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
