Assay Details
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012571.2 | NP_036703.2 | 4 - 5 | 626 | 69 | |
GenBank mRNA | BC061877.1 | - | 4 - 5 | 599 | 69 | |
CB578328.1 | - | 2 - 3 | 190 | 69 | ||
D00252.1 | - | 4 - 5 | 583 | 69 | ||
FM055803.1 | - | 4 - 5 | 548 | 69 | ||
J04171.1 | - | 4 - 5 | 567 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
