Assay Details
Gene Symbol: Pyroxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004261.1 | NP_001004261.1 | 12 - 13 | 1288 | 73 | |
XM_006231394.2 | XP_006231456.1 | 12 - 13 | 1559 | 73 | ||
XM_006231396.2 | XP_006231458.1 | 12 - 13 | 1560 | 73 | ||
XM_008760424.1 | XP_008758646.1 | 12 - 13 | 1562 | 73 | ||
XM_008760425.1 | XP_008758647.1 | 12 - 13 | 1325 | 73 | ||
GenBank mRNA | BC079368.1 | - | 12 - 13 | 1288 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
