Assay Details
Gene Symbol: Cacul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014248.1 | NP_001014270.1 | 6 - 7 | 1207 | 88 | |
XM_006231678.2 | XP_006231740.1 | 7 - 8 | 1457 | 88 | ||
GenBank mRNA | BC083839.1 | - | 6 - 7 | 1207 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
