Assay Details
Gene Symbol: Hoga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106355.1 | NP_001099825.1 | 1 - 2 | 408 | 96 | |
GenBank mRNA | FQ210513.1 | - | 1 - 2 | 222 | 96 | |
FQ218423.1 | - | 1 - 2 | 220 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
