Assay Details
Gene Symbol: Hoga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106355.1 | NP_001099825.1 | 3 - 4 | 676 | 57 | |
XM_006231386.2 | XP_006231448.1 | 2 - 3 | 674 | 57 | ||
GenBank mRNA | FQ210513.1 | - | 3 - 4 | 490 | 57 | |
FQ218423.1 | - | 3 - 4 | 488 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
