Assay Details
Gene Symbol: Actr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106364.1 | NP_001099834.1 | 2 - 3 | 182 | 72 | |
XM_006231470.2 | XP_006231532.1 | 2 - 3 | 294 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
