Assay Details
Gene Symbol: Kcnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033961.1 | NP_001029133.1 | 2 - 3 | 423 | 72 | |
NM_020094.2 | NP_064479.2 | 4 - 5 | 573 | 72 | ||
NM_020095.2 | NP_064480.2 | 3 - 4 | 519 | 72 | ||
XM_008760448.1 | XP_008758670.1 | 2 - 3 | 2131 | 72 | ||
XM_008760449.1 | XP_008758671.1 | 4 - 5 | 682 | 72 | ||
XM_008760450.1 | XP_008758672.1 | 2 - 3 | 323 | 72 | ||
GenBank mRNA | AF269283.1 | - | 4 - 5 | 351 | 72 | |
AF269284.1 | - | 3 - 4 | 297 | 72 | ||
AF269285.1 | - | 2 - 3 | 201 | 72 | ||
BC085905.1 | - | 2 - 3 | 423 | 72 | ||
GU937871.1 | - | 2 - 3 | 233 | 72 |
Gene Symbol: LOC100911951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231581.2 | XP_006231643.1 | 2 - 3 | 319 | 72 | |
XM_008760479.1 | XP_008758701.1 | 4 - 5 | 630 | 72 | ||
XM_008760480.1 | XP_008758702.1 | 3 - 4 | 575 | 72 | ||
XM_008760481.1 | XP_008758703.1 | 2 - 3 | 637 | 72 | ||
XM_008760482.1 | XP_008758704.1 | 2 - 3 | 478 | 72 | ||
XM_008760483.1 | XP_008758705.1 | 4 - 5 | 682 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
