Assay Details
Gene Symbol: Tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250448.2 | XP_006250510.1 | 1 - 2 | 1568 | 72 | |
GenBank mRNA | AF135598.1 | - | 1 - 2 | 352 | 72 | |
AF153013.1 | - | 1 - 2 | 658 | 72 | ||
AJ132718.1 | - | 1 - 2 | 366 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
