Assay Details
Gene Symbol: Disp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105983.1 | NP_001099453.1 | 7 - 8 | 1234 | 129 | |
XM_006250381.2 | XP_006250443.1 | 6 - 7 | 1103 | 129 | ||
XM_006250383.2 | XP_006250445.1 | 5 - 6 | 1564 | 129 | ||
XM_006250384.2 | XP_006250446.1 | 5 - 6 | 1043 | 129 | ||
XM_008769827.1 | XP_008768049.1 | 8 - 9 | 1928 | 129 | ||
XM_008769828.1 | XP_008768050.1 | 6 - 7 | 1214 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
