Assay Details
Gene Symbol: Disp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105983.1 | NP_001099453.1 | 3 - 4 | 423 | 52 | |
XM_008769827.1 | XP_008768049.1 | 4 - 5 | 1117 | 52 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001105983.1,XM_008769827.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
