Assay Details
Gene Symbol: Itpkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019312.2 | NP_062185.2 | 4 - 5 | 2704 | 83 | |
GenBank mRNA | AJ242781.1 | - | 2253 | 83 | ||
X74227.1 | - | 2027 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
