Assay Details
Gene Symbol: Ppox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105968.1 | NP_001099438.1 | 4 - 5 | 655 | 112 | |
XM_006250240.2 | XP_006250302.1 | 3 - 4 | 1556 | 112 | ||
XM_006250241.1 | XP_006250303.1 | 4 - 5 | 720 | 112 | ||
XM_006250243.1 | XP_006250305.1 | 3 - 4 | 363 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
