Assay Details
Gene Symbol: Ppox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105968.1 | NP_001099438.1 | 6 - 7 | 928 | 81 | |
XM_006250240.2 | XP_006250302.1 | 5 - 6 | 1829 | 81 | ||
XM_006250241.1 | XP_006250303.1 | 6 - 7 | 993 | 81 | ||
XM_006250243.1 | XP_006250305.1 | 5 - 6 | 636 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
