Assay Details
Gene Symbol: Rcor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134985.1 | NP_001128457.1 | 5 - 6 | 673 | 115 | |
GenBank mRNA | BC169033.1 | - | 5 - 6 | 673 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
