Assay Details
Gene Symbol: Ly9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191673.1 | NP_001178602.1 | 1 - 2 | 130 | 91 | |
XM_008769732.1 | XP_008767954.1 | 1 - 2 | 210 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
