Assay Details
Gene Symbol: Ly9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191673.1 | NP_001178602.1 | 4 - 5 | 1074 | 74 | |
XM_008769732.1 | XP_008767954.1 | 4 - 5 | 1154 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
