Assay Details
Gene Symbol: Ppp2r5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107891.1 | NP_001101361.1 | 12 - 13 | 1511 | 100 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P727824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173844 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P727824 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P727824 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P727824 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P142376 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P727824 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42503 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41010 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10770723 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770722 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770724 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770725 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770726 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10770721 | - | - |
Rat Genome 230 2.0 Array | 1392241_at | - | - |
Rat Genome 230 2.0 Array | 1392241_PM_at | - | - |
Rat Genome 230 2.0 Array | 1399005_at | - | - |
Rat Genome 230 2.0 Array | 1399005_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1399005_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1399005_at | - | - |
Affymetrix HT Rat Focus Array | 1392241_at | - | - |
Affymetrix HT Rat Focus Array | 1392241_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300015790.rn.1 | TC1300002218.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300015789.rn.1 | TC1300002218.rn.1 | RTA TC |
Rat Expression Set 230 | 1399005_PM_at | - | - |
Rat Expression Set 230 | 1399005_at | - | - |
Rat Expression Set 230 | 1392241_PM_at | - | - |
Rat Expression Set 230 | 1392241_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770721 | - | - |
Rat Genome U34 Set | rc_AA800651_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22282517 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370026 | - | - |
