Assay Details
Gene Symbol: Taf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037204.1 | NP_001032281.1 | 7 - 8 | 977 | 159 | |
XM_008769862.1 | XP_008768084.1 | 7 - 8 | 999 | 159 | ||
GenBank mRNA | BC107466.1 | - | 7 - 8 | 977 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
