Assay Details
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013079.1 | NP_001013097.1 | 2 - 3 | 198 | 134 | |
GenBank mRNA | BC091208.1 | - | 2 - 3 | 198 | 7381353 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC091208.1, NM_001013079.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
