Assay Details
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013079.1 | NP_001013097.1 | 3 - 4 | 341 | 111 | |
XM_006235816.2 | XP_006235878.1 | 4 - 5 | 590 | 111 | ||
GenBank mRNA | BC091208.1 | - | 3 - 4 | 341 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
