Assay Details
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013079.1 | NP_001013097.1 | 5 - 6 | 541 | 76 | |
XM_006235816.2 | XP_006235878.1 | 6 - 7 | 790 | 76 | ||
GenBank mRNA | BC091208.1 | - | 5 - 6 | 541 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
