Assay Details
Gene Symbol: Pcmtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107810.1 | NP_001101280.2 | 1 - 2 | 74 | 75 | |
GenBank mRNA | FQ211587.1 | - | 1 - 2 | 73 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107810.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
