Assay Details
Gene Symbol: Zgpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009656.1 | NP_001009656.1 | 5 - 6 | 1523 | 95 | |
XM_006235749.1 | XP_006235811.1 | 6 - 7 | 1519 | 95 | ||
XM_006235750.1 | XP_006235812.1 | 6 - 7 | 1484 | 95 | ||
XM_006235751.2 | XP_006235813.1 | 6 - 7 | 1505 | 95 | ||
XM_006235752.2 | XP_006235814.1 | 5 - 6 | 1419 | 95 | ||
GenBank mRNA | BC087720.1 | - | 5 - 6 | 1523 | 7318724 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
