Assay Details
Gene Symbol: Brat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751133.2 | XP_003751181.1 | 11 - 12 | 1666 | 81 | |
XM_006248968.2 | XP_006249030.1 | 11 - 12 | 1621 | 81 | ||
XM_006248972.1 | XP_006249034.1 | 12 - 13 | 1782 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
