Assay Details
Gene Symbol: Lnx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108329.1 | NP_001101799.1 | 8 - 9 | 1962 | 85 | |
XM_006248835.2 | XP_006248897.1 | 8 - 9 | 2089 | 85 | ||
XM_008768976.1 | XP_008767198.1 | 7 - 8 | 7008 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
