Assay Details
Gene Symbol: Pgm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106007.1 | NP_001099477.1 | 6 - 7 | 878 | 127 | |
XM_006250996.2 | XP_006251058.1 | 7 - 8 | 981 | 127 | ||
XM_006250997.2 | XP_006251059.1 | 8 - 9 | 1088 | 127 | ||
GenBank mRNA | BC160893.1 | - | 7 - 8 | 978 | 127 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106007.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
