Assay Details
Gene Symbol: Gabra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080587.3 | NP_542154.3 | 4 - 5 | 584 | 134 | |
XM_008770135.1 | XP_008768357.1 | 3 - 4 | 1271 | 134 | ||
GenBank mRNA | S55933.1 | - | 4 - 5 | 584 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
