Assay Details
Gene Symbol: Gabra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080587.3 | NP_542154.3 | 8 - 9 | 1220 | 124 | |
XM_008770135.1 | XP_008768357.1 | 7 - 8 | 1907 | 124 | ||
GenBank mRNA | S55933.1 | - | 8 - 9 | 1220 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20058 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15434 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15434 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15434 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15434 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89742 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48913 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86941 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90945 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10772529 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772523 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772524 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772528 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772531 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772530 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772527 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772526 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772525 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10772522 | - | - |
Rat Genome 230 2.0 Array | 1369905_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369905_at | - | - |
Affymetrix HT Rat Focus Array | 1369905_at | - | - |
Affymetrix HT Rat Focus Array | 1369905_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400003695.rn.1 | TC1400000465.rn.1 | RTA TC |
Rat Expression Set 230 | 1369905_at | - | - |
Rat Expression Set 230 | 1369905_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772522 | - | - |
Rat Genome U34 Set | S55933_i_at | - | - |
Rat Genome U34 Set | L08493cds_s_at | - | - |
Rat Genome U34 Set | S55933_i_at | - | - |
Rat Genome U34 Set | L08493cds_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22247297 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363867 | - | - |
RatRef-12 Expression BeadChip | ILMN_58901 | - | - |
