Assay Details
Gene Symbol: Rell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113776.1 | NP_001107248.1 | 5 - 6 | 754 | 83 | |
XM_006250998.2 | XP_006251060.1 | 5 - 6 | 849 | 83 | ||
GenBank mRNA | BC158872.1 | - | 5 - 6 | 754 | 83 | |
FQ224412.1 | - | 5 - 6 | 681 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P466321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43923 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P466321 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P466321 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P466321 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P466321 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10776957 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776955 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776956 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776958 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776959 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10776954 | - | - |
Rat Genome 230 2.0 Array | 1383357_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1383357_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400004221.rn.1 | TC1400000559.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400004222.rn.1 | TC1400000559.rn.1 | RTA TC |
Rat Expression Set 230 | 1383357_PM_a_at | - | - |
Rat Expression Set 230 | 1383357_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776954 | - | - |
Rat Genome U34 Set | rc_AI013497_f_at | - | - |
Rat Genome U34 Set | rc_AI009321_at | - | - |
Rat Genome U34 Set | rc_AI176632_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359256 | - | - |
