Assay Details
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250946.2 | XP_006251008.1 | 11 - 12 | 2120 | 144 | |
XM_006250947.2 | XP_006251009.1 | 11 - 12 | 2123 | 144 | ||
XM_006250948.2 | XP_006251010.1 | 11 - 12 | 2120 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
