Assay Details
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017452.1 | NP_001017452.1 | 2 - 3 | 603 | 119 | |
XM_006250946.2 | XP_006251008.1 | 2 - 3 | 894 | 119 | ||
XM_006250947.2 | XP_006251009.1 | 2 - 3 | 897 | 119 | ||
XM_006250948.2 | XP_006251010.1 | 2 - 3 | 894 | 119 | ||
GenBank mRNA | BC083653.1 | - | 2 - 3 | 603 | 7135159 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
