Assay Details
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250946.2 | XP_006251008.1 | 8 - 9 | 1777 | 181 | |
XM_006250947.2 | XP_006251009.1 | 8 - 9 | 1780 | 181 | ||
XM_006250948.2 | XP_006251010.1 | 8 - 9 | 1777 | 181 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
