Assay Details
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250946.2 | XP_006251008.1 | 10 - 11 | 1994 | 111 | |
XM_006250947.2 | XP_006251009.1 | 10 - 11 | 1997 | 111 | ||
XM_006250948.2 | XP_006251010.1 | 10 - 11 | 1994 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
