Assay Details
Gene Symbol: Lias | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012037.1 | NP_001012037.1 | 3 - 4 | 370 | 132 | |
XM_008770155.1 | XP_008768377.1 | 3 - 4 | 406 | 132 | ||
GenBank mRNA | BC083708.1 | - | 3 - 4 | 370 | 7190554 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
