Assay Details
Gene Symbol: Lias | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012037.1 | NP_001012037.1 | 9 - 10 | 1129 | 106 | |
GenBank mRNA | BC083708.1 | - | 9 - 10 | 1129 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
