Assay Details
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012956.1 | NP_037088.1 | 5 - 6 | 615 | 92 | |
XM_008770136.1 | XP_008768358.1 | 1 - 2 | 553 | 92 | ||
GenBank mRNA | X15466.1 | - | 5 - 6 | 615 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
