Assay Details
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012956.1 | NP_037088.1 | 6 - 7 | 756 | 166 | |
XM_008770136.1 | XP_008768358.1 | 2 - 3 | 694 | 166 | ||
GenBank mRNA | X15466.1 | - | 6 - 7 | 756 | 166 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
