Assay Details
Gene Symbol: Lnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108358.1 | NP_001101828.1 | 1 - 2 | 105 | 106 | |
XM_006250902.2 | XP_006250964.1 | 1 - 2 | 129 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108358.1, XM_006250902.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
