Assay Details
Gene Symbol: Lnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108358.1 | NP_001101828.1 | 2 - 3 | 576 | 85 | |
XM_006250902.2 | XP_006250964.1 | 2 - 3 | 600 | 85 | ||
GenBank mRNA | EU219404.1 | - | 1 - 2 | 419 | 85 | |
EU219405.1 | - | 1 - 2 | 419 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
