Assay Details
Gene Symbol: Lnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108358.1 | NP_001101828.1 | 3 - 4 | 821 | 55 | |
XM_006250902.2 | XP_006250964.1 | 3 - 4 | 845 | 55 | ||
XM_006250903.2 | XP_006250965.1 | 2 - 3 | 2853 | 55 | ||
XM_006250904.2 | XP_006250966.1 | 2 - 3 | 2853 | 55 | ||
GenBank mRNA | EU219404.1 | - | 2 - 3 | 664 | 55 | |
EU219405.1 | - | 2 - 3 | 664 | 55 | ||
EU219406.1 | - | 2 - 3 | 532 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
