Assay Details
Gene Symbol: Fip1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008295.1 | NP_001008296.1 | 12 - 13 | 1176 | 80 | |
XM_006250882.2 | XP_006250944.1 | 15 - 16 | 1484 | 80 | ||
XM_006250883.2 | XP_006250945.1 | 14 - 15 | 1457 | 80 | ||
XM_006250884.2 | XP_006250946.1 | 14 - 15 | 1413 | 80 | ||
XM_006250885.2 | XP_006250947.1 | 14 - 15 | 1374 | 80 | ||
XM_006250886.2 | XP_006250948.1 | 15 - 16 | 1486 | 80 | ||
XM_006250887.2 | XP_006250949.1 | 13 - 14 | 1303 | 80 | ||
GenBank mRNA | BC085767.1 | - | 12 - 13 | 1176 | 80 | |
EU224276.1 | - | 12 - 13 | 1175 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
