Assay Details
Gene Symbol: Uchl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017237.3 | NP_058933.2 | 6 - 7 | 512 | 119 | |
GenBank mRNA | BC060573.1 | - | 6 - 7 | 516 | 6920680 | 119 |
D10699.1 | - | 6 - 7 | 496 | 119 | ||
FQ214229.1 | - | 6 - 7 | 508 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
