Assay Details
Gene Symbol: Gnpda2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106005.1 | NP_001099475.1 | 1 - 2 | 90 | 129 | |
XM_006251036.2 | XP_006251098.1 | 1 - 2 | 140 | 129 | ||
GenBank mRNA | BC168866.1 | - | 1 - 2 | 77 | 7460205 | 129 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168866.1,NM_001106005.1,XM_006251036.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
