Assay Details
Gene Symbol: Gnpda2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106005.1 | NP_001099475.1 | 3 - 4 | 352 | 162 | |
XM_006251036.2 | XP_006251098.1 | 3 - 4 | 402 | 162 | ||
GenBank mRNA | BC168866.1 | - | 3 - 4 | 339 | 7460205 | 162 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
