Assay Details
Gene Symbol: Tmprss11d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033652.1 | NP_001028824.1 | 3 - 4 | 500 | 124 | |
GenBank mRNA | AF453776.1 | - | 3 - 4 | 500 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
