Assay Details
Gene Symbol: Sept11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107208.2 | NP_001100678.1 | 953 | 160 | ||
GenBank mRNA | BC167769.1 | - | 1084 | 160 | ||
EU711414.1 | - | 953 | 160 | |||
EU711415.1 | - | 953 | 160 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P058323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104367 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P058323 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P058323 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P058323 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P443030 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 429 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10775777 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775773 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775778 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775776 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775772 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775770 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775774 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775779 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775775 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775771 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10775769 | - | - |
Rat Genome 230 2.0 Array | 1372949_at | - | - |
Rat Genome 230 2.0 Array | 1396145_at | - | - |
Rat Genome 230 2.0 Array | 1382577_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372949_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382577_at | - | - |
Rat Genome 230 2.0 Array | 1396145_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396145_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382577_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396145_at | - | - |
Affymetrix HT Rat Focus Array | 1372949_at | - | - |
Affymetrix HT Rat Focus Array | 1372949_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382577_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400010781.rn.1 | TC1400001463.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400010791.rn.1 | TC1400001463.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400010780.rn.1 | TC1400001463.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400010784.rn.1 | TC1400001463.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400010789.rn.1 | TC1400001463.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400010790.rn.1 | TC1400001463.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400010779.rn.1 | TC1400001463.rn.1 | RTA TC |
Rat Expression Set 230 | 1372949_at | - | - |
Rat Expression Set 230 | 1372949_PM_at | - | - |
Rat Expression Set 230 | 1396145_PM_at | - | - |
Rat Expression Set 230 | 1382577_at | - | - |
Rat Expression Set 230 | 1396145_at | - | - |
Rat Expression Set 230 | 1382577_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775769 | - | - |
Rat Genome U34 Set | rc_AA818895_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374338 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040763 | - | - |
