Assay Details
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105998.1 | NP_001099468.1 | 4 - 5 | 720 | 104 | |
XM_006250715.2 | XP_006250777.1 | 3 - 4 | 820 | 104 | ||
GenBank mRNA | FQ223715.1 | - | 3 - 4 | 813 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
